α-Chloro-2,5-difluorotoulene 97%

Danger

H26735-14 H26735-06
AAH26735-14EA 521.7 USD
AAH26735-14 AAH26735-06
α-Chloro-2,5-difluorotoulene 97%
α-Chloro-2,5-difluorotoulene
Formula: C₇H₅ClF₂
MW: 162.57 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01090989
CAS Number: 495-07-8
EINECS: None
UN: 3261
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR