2-Chloro-6-fluoro-m-cresol 97%

Danger

Synonyms: 2-Chloro-6-fluoro-3-methylphenol2-Choro-6-fluoro-3-methylphenol

H26721-14 H26721-06 H26721-03
AAH26721-14EA 560.55 USD
AAH26721-14 AAH26721-03 AAH26721-06
2-Chloro-6-fluoro-m-cresol 97%
2-Chloro-6-fluoro-m-cresol
Formula: C₇H₆ClFO
MW: 160.58 g/mol
Boiling Pt: 211 °C
Melting Pt: 47…50 °C
Storage Temperature: Ambient
MDL Number: MFCD01631366
CAS Number: 261762-90-7
UN: 3261
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR