4-Chloro-2-(trifluoromethyl)benzonitrile 97%

Warning

H26675-06 H26675-03
AAH26675-06EA 210.9 USD
AAH26675-06 AAH26675-03
4-Chloro-2-(trifluoromethyl)benzonitrile 97%
4-Chloro-2-(trifluoromethyl)benzonitrile
Formula: C₈H₃ClF₃N
MW: 205.57 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00060671
CAS Number: 320-41-2
UN: 3276
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR