4-Fluoro-m-toluonitrile 97%

Warning

H26658-03 H26658-09
AAH26658-09EA 351.87 USD
AAH26658-09 AAH26658-03
4-Fluoro-m-toluonitrile 97%
4-Fluoro-m-toluonitrile
Formula: C₈H₆FN
MW: 135.14 g/mol
Melting Pt: 53…56 °C
Storage Temperature: Ambient
MDL Number: MFCD03788510
CAS Number: 185147-08-4
EINECS: None
UN: 3439
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR