4-Fluoro-o-toluonitrile 97%

Supplier: Thermo Scientific Chemicals
Warning

H26641-06 H26641-14
AAH26641-14EA 456.99 USD
AAH26641-14 AAH26641-06
4-Fluoro-o-toluonitrile 97%
4-Fluoro-o-toluonitrile
Formula: C₈H₆FN
MW: 135.14 g/mol
Melting Pt: 67…71 °C
Storage Temperature: Ambient
MDL Number: MFCD03095106
CAS Number: 147754-12-9
UN: 3439
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR