2,4-Difluoro-3-methoxybenzyl bromide 97%

Danger

H26625-03 H26625-06
AAH26625-03EA 126.12 USD
AAH26625-03 AAH26625-06
2,4-Difluoro-3-methoxybenzyl bromide 97%
2,4-Difluoro-3-methoxybenzyl bromide
Formula: C₈H₇BrF₂O
Storage Temperature: Ambient
MDL Number: MFCD04115936
CAS Number: 886499-17-8
UN: 3265
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR