3,5-Difluoro-2-methoxybenzyl bromide 97%

Danger

Synonyms: 1-(Bromomethyl)-3,5-difluoro-2-methoxybenzene

H26582-06 H26582-03
AAH26582-03EA 151.95 USD
AAH26582-03 AAH26582-06
3,5-Difluoro-2-methoxybenzyl bromide 97%
3,5-Difluoro-2-methoxybenzyl bromide
Formula: C₈H₇BrF₂O
MW: 237.04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06660240
CAS Number: 886500-63-6
UN: 3265
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR