2,6-Difluoro-4-methoxybenzyl bromide 97%

Danger

Synonyms: 2-(Bromomethyl)-1,3-difluoro-5-methoxybenzene

H26576-06 H26576-03
AAH26576-06EA 595.43 USD
AAH26576-06 AAH26576-03
2,6-Difluoro-4-methoxybenzyl bromide 97%
2,6-Difluoro-4-methoxybenzyl bromide
Formula: C₈H₇BrF₂O
MW: 237.04 g/mol
Melting Pt: 43…46 °C
Storage Temperature: Ambient
MDL Number: MFCD04115925
CAS Number: 94278-68-9
UN: 3261
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR