2-Methyl-5-(trifluoromethyl)benzonitrile 97%

Danger

H26549-09 H26549-03
AAH26549-09EA 434.01 USD
AAH26549-09 AAH26549-03
2-Methyl-5-(trifluoromethyl)benzonitrile 97%
2-Methyl-5-(trifluoromethyl)benzonitrile
Formula: C₉H₆F₃N
Storage Temperature: Ambient
MDL Number: MFCD01631594
CAS Number: 261952-03-8
UN: 3276
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR