2,4-Difluoro-3-methoxyaniline 97%

Supplier: Thermo Scientific Chemicals
Danger

H26489-06
AAH26489-06EA 623.99 USD
AAH26489-06
2,4-Difluoro-3-methoxyaniline 97%
2,4-Difluoro-3-methoxyaniline
Formula: C₇H₇F₂NO
Storage Temperature: Ambient
MDL Number: MFCD04115929
CAS Number: 886499-08-7
UN: 2810
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR