2,3,5-Trifluorobenzylamine 97%

Danger

Synonyms: (2,3,5-Trifluorophenyl)methanamine

H26469-03 H26469-06
AAH26469-03EA 140.97 USD
AAH26469-03 AAH26469-06
2,3,5-Trifluorobenzylamine 97%
2,3,5-Trifluorobenzylamine
Formula: C₇H₆F₃N
MW: 161.13 g/mol
Boiling Pt: 73 °C/3 mm
Storage Temperature: Ambient
MDL Number: MFCD00236314
CAS Number: 244022-72-8
EINECS: None
UN: 2735
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR