2,3-Difluoro-4-methoxybenzyl bromide 97%

Danger

H26434-03 H26434-06
AAH26434-03EA 110.55 USD
AAH26434-03 AAH26434-06
2,3-Difluoro-4-methoxybenzyl bromide 97%
2,3-Difluoro-4-methoxybenzyl bromide
Formula: C₈H₇BrF₂O
Storage Temperature: Ambient
MDL Number: MFCD04115985
CAS Number: 689254-23-7
UN: 3261
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR