3-Fluoro-o-toluonitrile 97%

Warning

H26413-03 H26413-06
AAH26413-03EA 54.46 USD
AAH26413-03 AAH26413-06
3-Fluoro-o-toluonitrile 97%
3-Fluoro-o-toluonitrile
Formula: C₈H₆FN
MW: 135.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03788541
CAS Number: 185147-06-2
UN: 3276
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR