2,3-Difluoro-6-methoxybenzyl bromide 97%

Danger

H26380-03 H26380-06
AAH26380-06EA 228.66 USD
AAH26380-06 AAH26380-03
2,3-Difluoro-6-methoxybenzyl bromide 97%
2,3-Difluoro-6-methoxybenzyl bromide
Formula: C₈H₇BrF₂O
Melting Pt: 46…48 °C
Storage Temperature: Ambient
MDL Number: MFCD04116023
CAS Number: 886501-83-3
UN: 3261
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR