α-Bromo-3-chloro-5-fluorotoluene 97%

Danger

Synonyms: 1-(Bromomethyl)-3-chloro-5-fluorobenzene3-Chloro-5-fluorobenzyl bromide

H26309-06 H26309-03
AAH26309-06EA 333 USD
AAH26309-06 AAH26309-03
α-Bromo-3-chloro-5-fluorotoluene 97%
α-Bromo-3-chloro-5-fluorotoluene
Formula: C₇H₅BrClF
MW: 223.47 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03788472
CAS Number: 493024-39-8
UN: 3265
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR