2-Fluoro-4-hydroxybenzaldehyde 97%

Supplier: Thermo Scientific Chemicals

Warning

H26218-ME H26218-MC
AAH26218-MCEA 110 USD
AAH26218-MC AAH26218-ME
2-Fluoro-4-hydroxybenzaldehyde 97%
2-Fluoro-4-hydroxybenzaldehyde
Formula: C₇H₅FO₂
MW: 140.11 g/mol
Melting Pt: 168…170 °C
Storage Temperature: Ambient
MDL Number: MFCD06797918
CAS Number: 348-27-6
EINECS: None

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR