2-Fluoro-5-iodobenzonitrile 97%

Warning

H26172-06 H26172-03
AAH26172-03EA 49.34 USD
AAH26172-03 AAH26172-06
2-Fluoro-5-iodobenzonitrile 97%
2-Fluoro-5-iodobenzonitrile
Formula: C₇H₃FIN
MW: 247.01 g/mol
Melting Pt: 68…72 °C
Storage Temperature: Ambient
MDL Number: MFCD03094225
CAS Number: 351003-36-6
UN: 3439
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR