2-Methoxy-5-(trifluoromethyl)benzaldehyde 98%

Supplier: Thermo Scientific Chemicals
Warning

H26135-MD H26135-03
AAH26135-MDEA 80.93 USD
AAH26135-MD AAH26135-03
2-Methoxy-5-(trifluoromethyl)benzaldehyde 98%
2-Methoxy-5-(trifluoromethyl)benzaldehyde
Formula: C₉H₇F₃O₂
MW: 204.15 g/mol
Melting Pt: 85…89 °C
Storage Temperature: Ambient
MDL Number: MFCD03788528
CAS Number: 146539-83-5
EINECS: None

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR