6-Fluoropicolinic acid 97%

Warning

Synonyms: 2-Fluoro-6-pyridinecarboxylic acid6-Fluoropyridine-2-carboxylic acid

H26106-MD H26106-03
AAH26106-MDEA 115.68 USD
AAH26106-MD AAH26106-03
6-Fluoropicolinic acid 97%
6-Fluoropicolinic acid
Formula: C₆H₄FNO₂
MW: 141.1 g/mol
Melting Pt: 139…143 °C
Storage Temperature: Ambient
MDL Number: MFCD02181193
CAS Number: 402-69-7
EINECS: None

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR