trans-2-Fluorocyclohexanol 97%

Warning

H26031-03
AAH26031-03EA 134.31 USD
AAH26031-03
trans-2-Fluorocyclohexanol 97%
trans-2-Fluorocyclohexanol
Formula: C₆H₁₁FO
MW: 118.15 g/mol
Boiling Pt: 166 °C
Melting Pt: 21 °C
Storage Temperature: Ambient
MDL Number: MFCD08146648
CAS Number: 656-60-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR