6-Chloro-2-fluoropurine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 6-Chloro-2-fluoro-9H-purine

H25825-ME
AAH25825-MEEA 197.99 USD
AAH25825-ME
6-Chloro-2-fluoropurine 97%
6-Chloro-2-fluoropurine
Formula: C₅H₂ClFN₄
MW: 172.55 g/mol
Melting Pt: 151…153 °C
Storage Temperature: Ambient
MDL Number: MFCD02183557
CAS Number: 1651-29-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR