6-Bromo-1,4-benzodioxane 98%

Warning

Synonyms: 6-Bromo-2,3-dihydro-1,4-benzodioxine6-Bromo-2,3-dihydrobenzo[b][1,4]dioxine

B25625-06 B25625-03 B25625-03
AAAB25625-03EA 27.72 USD
AAAB25625-03 CAAAAB25625-03 CAAAAB25625-06
6-Bromo-1,4-benzodioxane 98%
6-Bromo-1,4-benzodioxane
Formula: C₈H₇BrO₂
MW: 215.05 g/mol
Boiling Pt: 259…260 °C
Density: 1.598
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00040750
CAS Number: 52287-51-1
EINECS: 257-817-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR