4-Chloro-4'-hydroxybutyrophenone 95%

Warning

Synonyms: 4-Chloro-1-(4-hydroxyphenyl)butan-1-one

B25543-06 B25543-14 B25543-14
AAAB25543-14EA 208.68 USD
CAAAAB25543-06 AAAB25543-14 CAAAAB25543-14
4-Chloro-4'-hydroxybutyrophenone 95%
4-Chloro-4'-hydroxybutyrophenone
Formula: C₁₀H₁₁ClO₂
MW: 198.65 g/mol
Melting Pt: 105…110 °C
Storage Temperature: Ambient
MDL Number: MFCD00002362
CAS Number: 7150-55-2
EINECS: 230-485-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR