4-Fluoro-1-naphthonitrile 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Fluoronaphthalene-1-carbonitrile
B25335-14
B25335-06
B25335-03
AAB25335-06EA
136.99
USD
AAB25335-06
AAB25335-14
AAB25335-03
4-Fluoro-1-naphthonitrile 97%
4-Fluoro-1-naphthonitrile
Formula:
C₁₁H₆FN MW: 171.18 g/mol Melting Pt: 89…91 °C Storage Temperature: Ambient |
MDL Number:
MFCD00236656 CAS Number: 13916-99-9 UN: 3439 ADR: "6.1",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...