Methyl-4-aminonicotinate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl 4-aminonicotinate

B25324-03 B25324-06
AAB25324-03EA 36.24 USD
AAB25324-03 AAB25324-06
Methyl-4-aminonicotinate 98%
Methyl 4-aminonicotinate
Formula: C₇H₈N₂O₂
MW: 152.15 g/mol
Melting Pt: 172…174 °C
Storage Temperature: Ambient
MDL Number: MFCD00955996
CAS Number: 16135-36-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR