Ethyl-2,6-dichlorobenzoate 97%

Warning

B25275-06 B25275-03 B25275-06 B25275-14 B25275-14 B25275-03
AAAB25275-03EA 50.1 USD
AAAB25275-03 AAAB25275-14 AAAB25275-06 CAAAAB25275-06 CAAAAB25275-14 CAAAAB25275-03
Ethyl-2,6-dichlorobenzoate 97%
Ethyl 2,6-dichlorobenzoate
Formula: C₉H₈Cl₂O₂
MW: 219.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01313702
CAS Number: 81055-73-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR