4-Phenylazophenol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-(Phenyldiazenyl)phenol
B25198-14
B25198-06
B25198-22
B25198-14
B25198-06
B25198-22
AAAB25198-06EA
20.09
USD
AAAB25198-06
CAAAAB25198-06
AAAB25198-14
CAAAAB25198-14
AAAB25198-22
CAAAAB25198-22
4-Phenylazophenol 98%
4-Phenylazophenol
Formula:
C₁₂H₁₀N₂O MW: 198.23 g/mol Melting Pt: 152…155 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002330 CAS Number: 1689-82-3 EINECS: 216-880-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...