1-Methyl-1-(2-nitro-4-(trifluoromethyl)phenyl)hydrazine 97%

Warning

B24823-01 B24823-03 B24823-01
AAB24823-01EA 98.56 USD
CAAAB24823-01 CAAAB24823-03 AAB24823-01
1-Methyl-1-(2-nitro-4-(trifluoromethyl)phenyl)hydrazine 97%
1-Methyl-1-(2-nitro-4-(trifluoromethyl)phenyl)hydrazine
Formula: C₈H₈F₃N₃O₂
MW: 235.16
Melting Pt: 64…67 °C
Storage Temperature: Ambient
MDL Number: MFCD00052691
CAS Number: 120940-43-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR