2-Amino-4-methoxy-6-methylpyrimidine 99%

Warning

B24673-06 B24673-14 B24673-14 B24673-06
AAB24673-06EA 440 USD
CAAAB24673-14 CAAAB24673-06 AAB24673-06 AAB24673-14
2-Amino-4-methoxy-6-methylpyrimidine 99%
2-Amino-4-methoxy-6-methylpyrimidine
Formula: C₆H₉N₃O
Melting Pt: 158…164 °C
Storage Temperature: Ambient
MDL Number: MFCD00059768
CAS Number: 7749-47-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR