Phenylphosphonic acid 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Phenylphosphonic Acid

B22939-36 B22939-22 B22939-36 B22939-22
AAAB22939-36EA 293.26 USD
AAAB22939-36 CAAAAB22939-36 AAAB22939-22 CAAAAB22939-22
Phenylphosphonic acid 98%
Phenylphosphonic acid
Formula: C₆H₇O₃P
MW: 158.09 g/mol
Melting Pt: 160…162 °C
Storage Temperature: Ambient
MDL Number: MFCD00002136
CAS Number: 1571-33-1
EINECS: 216-388-1
UN: 3261
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR