4-Chlorophenyl cyclopropyl ketone 98%

Synonyms: (4-Chlorophenyl)(cyclopropyl)methanone

B22766-09 B22766-30 B22766-18 B22766-30 B22766-18 B22766-09
AAB22766-18EA 171.23 USD
CAAAB22766-18 CAAAB22766-09 AAB22766-18 CAAAB22766-30 AAB22766-09 AAB22766-30
4-Chlorophenyl cyclopropyl ketone 98%
4-Chlorophenyl cyclopropyl ketone
Formula: C₁₀H₉ClO
MW: 180.63 g/mol
Boiling Pt: 135…137 °C/10 mm
Melting Pt: 29…31 °C
Density: 1.160
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001295
CAS Number: 6640-25-1
EINECS: 229-655-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR