Benzoin isopropyl ether 98+%

Synonyms: 2-Isopropoxy-1,2-diphenylethanone

B22654-22 B22654-14 B22654-22 B22654-14 B22654-36 B22654-36
AAB22654-14EA 195.1 USD
CAAAB22654-22 CAAAB22654-14 CAAAB22654-36 AAB22654-14 AAB22654-36 AAB22654-22
Benzoin isopropyl ether 98+%
Benzoin isopropyl ether
Formula: C₁₇H₁₈O₂
MW: 254.33 g/mol
Boiling Pt: 180 °C/10 mm
Melting Pt: 76…79 °C
Storage Temperature: Ambient
MDL Number: MFCD00015025
CAS Number: 6652-28-4
EINECS: 229-677-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR