7-Methoxy-2-tetralone 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 3,4-Dihydro-7-methoxynaphthalen-2(1H)-one7-Methoxy-3,4-dihydronaphthalen-2(1H)-one

B22432-06 B22432-03 B22432-03 B22432-06
AAB22432-06EA 129.99 USD
CAAAB22432-06 AAB22432-06 CAAAB22432-03 AAB22432-03
7-Methoxy-2-tetralone 98%
7-Methoxy-2-tetralone
Formula: C₁₁H₁₂O₂
MW: 176.22 g/mol
Boiling Pt: 124…126 °C/1.5 mm
Density: 1.130
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001730
CAS Number: 4133-34-0
EINECS: 223-954-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR