4,4''-Dinitro-p-terphenyl 98+%

Warning

B22059-14 B22059-14 B22059-06 B22059-06
AAAB22059-06EA 115.44 USD
AAAB22059-06 CAAAAB22059-06 AAAB22059-14 CAAAAB22059-14
4,4''-Dinitro-p-terphenyl 98+%
4,4''-Dinitro-p-terphenyl
Formula: C₁₈H₁₂N₂O₄
Melting Pt: 276…280 °C
Storage Temperature: Ambient
MDL Number: MFCD00051743
CAS Number: 3282-11-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR