1-Chloro-4-(trifluoromethoxy)benzene 98%

Synonyms: 4-(Trifluoromethoxy)chlorobenzene

B21437-09 B21437-09 B21437-04 B21437-04
AAB21437-09EA 128.99 USD
CAAAB21437-09 AAB21437-09 AAB21437-04 CAAAB21437-04
1-Chloro-4-(trifluoromethoxy)benzene 98%
1-Chloro-4-(trifluoromethoxy)benzene
Formula: C₇H₄ClF₃O
MW: 196.56 g/mol
Boiling Pt: 142…143 °C
Density: 1,37
Storage Temperature: Ambient
MDL Number: MFCD00040953
CAS Number: 461-81-4
EINECS: 207-315-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR