cis-1,2,3,6-Tetrahydrophthalic anhydride 95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: cis-4-Cyclohexene-1,2-dicarboxylic anhydride

B21431-30 B21431-30
AAB21431-30EA 29.91 USD
AAB21431-30 CAAAB21431-30
cis-1,2,3,6-Tetrahydrophthalic anhydride 95%
cis-1,2,3,6-Tetrahydrophthalic anhydride
Formula: C₈H₈O₃
Melting Pt: 97…103 °C
Storage Temperature: Ambient
MDL Number: MFCD00005916
CAS Number: 935-79-5
EINECS: 213-308-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR