2,5-Dichlorohydroquinone 97%

Supplier: Thermo Scientific Chemicals
Danger

B21285-06 B21285-03 B21285-03 B21285-06
AAB21285-03EA 50.29 USD
CAAAB21285-03 CAAAB21285-06 AAB21285-03 AAB21285-06
2,5-Dichlorohydroquinone 97%
2,5-Dichlorohydroquinone
Formula: C₆H₄Cl₂O₂
Melting Pt: 168…171 °C
Storage Temperature: Ambient
MDL Number: MFCD00041749
CAS Number: 824-69-1
EINECS: 212-533-8
UN: 1759
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR