Methyl-5-bromonicotinate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Methyl 5-bromonicotinate
B20908-03
B20908-06
B20908-06
B20908-03
AAB20908-06EA
115.99
USD
AAB20908-06
AAB20908-03
CAAAB20908-03
CAAAB20908-06
Methyl-5-bromonicotinate 97%
Methyl 5-bromonicotinate
Formula:
C₇H₆BrNO₂ MW: 216.03 g/mol Melting Pt: 96…98 °C Storage Temperature: Ambient |
MDL Number:
MFCD00173668 CAS Number: 29681-44-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...