2,6-Dichloroisonicotinic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,6-Dichloropyridine-4-carboxylic acid

B20541-06 B20541-03
AAAB20541-06EA 142.99 USD
AAAB20541-06 AAAB20541-03
2,6-Dichloroisonicotinic acid 98%
2,6-Dichloroisonicotinic acid
Formula: C₆H₃Cl₂NO₂
Melting Pt: 208 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00234147
CAS Number: 5398-44-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR