2-Amino-3,5-dibromo-6-methylpyridine 97%

Warning

Synonyms: 3,5-Dibromo-6-methyl-2-pyridinamine

B20498-06 B20498-03 B20498-06 B20498-03
AAB20498-03EA 54.78 USD
CAAAB20498-06 AAB20498-03 AAB20498-06 CAAAB20498-03
2-Amino-3,5-dibromo-6-methylpyridine 97%
2-Amino-3,5-dibromo-6-methylpyridine
Formula: C₆H₆Br₂N₂
MW: 265.94 g/mol
Melting Pt: 144…145 °C
Storage Temperature: Ambient
MDL Number: MFCD00068229
CAS Number: 91872-10-5
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR