3',5'-Difluoro-4'-hydroxypropiophenone 98%

Warning

B20229-03 B20229-06 B20229-03 B20229-06
AAB20229-06EA 295.26 USD
AAB20229-06 AAB20229-03 CAAAB20229-06 CAAAB20229-03
3',5'-Difluoro-4'-hydroxypropiophenone 98%
3',5'-Difluoro-4'-hydroxypropiophenone
Formula: C₉H₈F₂O₂
Melting Pt: 128…132 °C
Storage Temperature: Ambient
MDL Number: MFCD00061287
CAS Number: 178374-78-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR