2',5'-Diethoxybenzanilide 97%

B20227-06 B20227-14 B20227-03 B20227-14 B20227-06 B20227-03
AAB20227-06EA 112.57 USD
AAB20227-06 AAB20227-14 AAB20227-03 CAAAB20227-14 CAAAB20227-03 CAAAB20227-06
2',5'-Diethoxybenzanilide 97%
2',5'-Diethoxybenzanilide
Formula: C₁₇H₁₉NO₃
Melting Pt: 87…89 °C
Storage Temperature: Ambient
MDL Number: MFCD00026798
CAS Number: 92-22-8
EINECS: 202-137-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR