Cyclopentane-1,1-diacetic acid 98+%

Warning

Synonyms: 2,2-(Cyclopentane-1,1-diyl)diacetic acid3,3-Tetramethyleneglutaric acid

B20146-22 B20146-22 B20146-14 B20146-14
AAB20146-14EA 82.27 USD
CAAAB20146-14 CAAAB20146-22 AAB20146-14 AAB20146-22
Cyclopentane-1,1-diacetic acid 98+%
Cyclopentane-1,1-diacetic acid
Formula: C₉H₁₄O₄
MW: 186.21 g/mol
Melting Pt: 179…181 °C
Storage Temperature: Ambient
MDL Number: MFCD00001388
CAS Number: 16713-66-9
EINECS: 240-761-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR