Nitrofen 98%

Supplier: Thermo Scientific Chemicals

Danger

Synonyms: 2,4-Dichloro-1-(4-nitrophenoxy)benzene

A19977-14 A19977-14
AAA19977-14EA 271 USD
AAA19977-14 CAAAA19977-14
Nitrofen 98%
Nitrofen
Formula: C₁₂H₇Cl₂NO₃
MW: 284.1 g/mol
Melting Pt: 68…71 °C
Storage Temperature: Ambient
MDL Number: MFCD00128026
CAS Number: 1836-75-5
EINECS: 217-406-0
UN: 3077
ADR: "9",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR