(4-Methylphenoxy)acetonitrile 97%

Supplier: Alfa Aesar
Danger

A19926-06 A19926-14 A19926-06 A19926-14
AAAA19926-14EA 470.5 USD
CAAAA19926-06 AAAA19926-14 AAA19926-06 CAAAAA19926-14
(4-Methylphenoxy)acetonitrile 97%
(4-Methylphenoxy)acetonitrile
Formula: C₉H₉NO
Boiling Pt: 76…78 °C (11 mmHg)
Melting Pt: 42…44 °C
Storage Temperature: Ambient
MDL Number: MFCD00068121
CAS Number: 33901-44-9
EINECS: 000-000-0
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR