Methyl-3,4-difluorobenzoate 97%

Warning

Synonyms: Methyl 3,4-difluorobenzoate

A19866-06 A19866-03 A19866-03 A19866-06
AAA19866-06EA 87.37 USD
AAA19866-06 AAA19866-03 CAAAA19866-03 CAAAA19866-06
Methyl-3,4-difluorobenzoate 97%
Methyl 3,4-difluorobenzoate
Formula: C₈H₆F₂O₂
MW: 172.13 g/mol
Boiling Pt: 87…89 °C/15 mm
Storage Temperature: Ambient
MDL Number: MFCD00276597
CAS Number: 369-25-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR