Hydrazinium monobromide 98%

Danger

Synonyms: Hydrazine monohydrobromide

A19462-30 A19462-30
AAA19462-30EA 147.08 USD
CAAAA19462-30 AAA19462-30
Hydrazinium monobromide 98%
Hydrazinium monobromide

Special handling precautions required. View MSDS prior to purchase.

Formula: H₅BrN₂
Boiling Pt: ca 190 °C (dec.)
Melting Pt: 92…93 °C
Storage Temperature: Ambient
MDL Number: MFCD00065394
CAS Number: 13775-80-9
EINECS: 237-412-7
UN: 2928
ADR: "6.1",II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR