Cyclopentadecane 98%

Supplier: Thermo Scientific Chemicals

A19416-03 A19416-03 A19416-06 A19416-06
AAA19416-06EA 152 USD
CAAAA19416-03 CAAAA19416-06 AAA19416-06 AAA19416-03
Cyclopentadecane 98%
Cyclopentadecane
Formula: C₁₅H₃₀
Density: 0,805
Storage Temperature: Ambient
MDL Number: MFCD00039424
CAS Number: 295-48-7
EINECS: 206-040-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR