3-Amino-4-methoxybenzanilide 98%

A19168-18 A19168-09 A19168-18 A19168-09
AAA19168-09EA 148.16 USD
CAAAA19168-09 CAAAA19168-18 AAA19168-09 AAA19168-18
3-Amino-4-methoxybenzanilide 98%
3-Amino-4-methoxybenzanilide
Formula: C₁₄H₁₄N₂O₂
MW: 242.28 g/mol
Melting Pt: 154…156 °C
Storage Temperature: Ambient
MDL Number: MFCD00017166
CAS Number: 120-35-4
EINECS: 204-388-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR