p-Xylylenebis(triphenylphosphonium bromide) 96%

Supplier: Thermo Scientific Chemicals
Warning

A18241-14 A18241-22 A18241-14 A18241-22
AAA18241-22EA 129.99 USD
CAAAA18241-22 CAAAA18241-14 AAA18241-22 AAA18241-14
p-Xylylenebis(triphenylphosphonium bromide) 96%
p-Xylylenebis(triphenylphosphonium bromide)
Formula: C₄₄H₃₈Br₂P₂
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00011914
CAS Number: 40817-03-6
EINECS: 255-092-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR